Order Entry
Ireland
ContactUsLinkComponent
3-Ethyl-3-hexanol ≥98%
Irritant
3-Ethyl-3-hexanol ≥98%
Catalog # B20859.06
CAS Number:  
Irritant
3-Ethyl-3-hexanol ≥98%
Catalog # B20859.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Density:0,83
 :130.23 g/mol
EINECS:209-908-3
 :GHS07
Storage Temperature:Ambient
CAS Number:597-76-2
Formula:C₈H₁₈O
MDL Number:MFCD00039616