

Acetonitrile
Catalog # ROTH2259.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
About this item
:
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:81,6 °C (1013 hPa)
Melting Pt:–45,7 °C
Density:0,782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS02
:GHS07
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878