Order Entry
Ireland
ContactUsLinkComponent
2,4,6-Tris(dimethylaminomethyl)phenol, Sigma-Aldrich®
2,4,6-Tris(dimethylaminomethyl)phenol, Sigma-Aldrich®
Catalog # 45348-250ML-F
CAS Number:  
undefined
2,4,6-Tris(dimethylaminomethyl)phenol, Sigma-Aldrich®
Catalog # 45348-250ML-F
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :265.4 g/mol
Boiling Pt:130…135 °C (1 mmHg)
Density:0,974 g/cm³ (15 °C)
EINECS:202-013-9
CAS Number:90-72-2
Formula:C₁₅H₂₇N₃O
MDL Number:MFCD00008330