Order Entry
Ireland
ContactUsLinkComponent
 
Flammable
Toxic
Corrosive
HMDS (1,1,1,3,3,3-hexamethyldisilazane)
Catalog # HYPETS-84770
CAS Number:  
undefined
Flammable
Toxic
Corrosive
HMDS (1,1,1,3,3,3-hexamethyldisilazane)
Catalog # HYPETS-84770
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :161.39 g/mol
Merck Index:13,04708
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–82 °C
Density:0,7742 g/cm³ (20 °C)
Flash Pt:6 °C
Storage Temperature:Ambient
EINECS:213-668-5
UN:2924
ADR:3,II
REACH:01-2119438176-38
 :GHS02
 :GHS06
 :GHS05
CAS Number:999-97-3
Formula:(CH₃)₃SiNHSi(CH₃)₃
MDL Number:MFCD00008259