Tris(perfluorobutyl)amine tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine
CAS Number:311-89-7
:
Formula:C₁₂F₂₇N
MDL Number:MFCD00000436
Boiling Pt:177…180 °C
Melting Pt:–52 °C
Density:1.880
Molecular Weight:671.1 g/mol
Storage Temperature:Ambient
EINECS:206-223-1
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together