Dopamine hydrochloride 99%
:62-31-7
:
:C₈H₁₂ClNO₂
:MFCD00012898
:189.64
:241…243 °C
:1,356 g/cm³ (20 °C)
:Ambient
:200-527-8
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (105°C, 2 hrs)
- Sulfated ash:≤0.1 %
- Titration with HClO4:≥98.5 % (On dried substance)
- Heavy metals (as Pb):≤20 ppm
Frequently Bought Together