Order Entry
Ireland
ContactUsLinkComponent
Triphosgene, Sigma-Aldrich®
Toxic
Triphosgene, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Triphosgene, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    77 - 81 °C
  • Molar Mass:
    296.75 g/mol
  • Boiling Point:
    105 °C (30 hPa)
  • Vapor Pressure:
    16 hPa (90 °C)
  • Density:
    1.6 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together