
(1R,2R)-(-)-1,2-Cyclohexanedicarboxylic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:46022-05-3
:
Formula:C₈H₁₂O₄
MDL Number:MFCD00062973
:172.18 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together