Order Entry
Ireland
ContactUsLinkComponent
VWR® CERTICLEAN® Cleanroom Gloves, Latex
VWR® CERTICLEAN® Cleanroom Gloves, Latex
  • Catalog No:
  • KIMB40101-052_INT
  • KIMB40101-044_INT
  • KIMB40101-056_INT
  • KIMB40101-048_INT
  • KIMB40101-058_INT
  • Model:
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • Cuff style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Finger thickness:
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • 0,22 mm
  • Palm thickness:
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
  • 0,20 mm
Frequently Bought Together