Order Entry
Ireland
ContactUsLinkComponent
Pinch clamp, Nalgene®
Pinch clamp, Nalgene®
Catalog # NALG6165-0002
   Low Manufacturing Impact
undefined
Pinch clamp, Nalgene®
Catalog # NALG6165-0002
   Low Manufacturing Impact
Specifications
  • Spanning width:
    6 - 11 mm
  • Colour:
    White
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    12
Specifications
About this item
This pinch clamp made of PP enables precise, one-hand regulation of fluid flow - from completely open to restricted flow to completely closed.
  • Plastic clamp will not rust, corrode or deform
  • Molded in one piece with no sharp edges
  • Accommodates a wide range of tubing