
Caprinaldehyde stabilised for synthesis, Sigma-Aldrich®
Catalog # 8.03211.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 ml
- Seq:2440
Specifications
Specification Test Results
- Hill Formula:C₁₀H₂₀O
- Chemical formula:CH₃(CH₂)₈CHO
- Molar Mass:156.26 g/mol
- Boiling point:207 - 209 °C (1013 hPa)
- Density:0.83 g/cm³ (20 °C)
- Flash point:87 °C
- Ignition temperature:200 °C
- Melting Point:7 °C
- Vapor pressure:0.1 hPa (20 °C)
- Refractive Index:1.428 (20 °C, 589 nm)
- Solubility:(20 °C) practically insoluble
- LD 50 oral:LD50 Rat 3096 mg/kg
- LD 50 dermal:LD50 Rabbit 4183 mg/kg
- Storage class:10 Combustible liquids not in Storage Class 3
- WGK:WGK 2 water endangering
- R Phrase:R 38
Specification Test Results
About this item
:
:156.27 g/mol
Boiling Pt:207 °C (1013 hPa)
Melting Pt:7 °C
Density:0,83 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:Stabilised with α-tocopherol
Storage Temperature:Ambient
EINECS:203-957-4
:GHS07
CAS Number:112-31-2
Formula:C₁₀H₂₀O
MDL Number:MFCD00007031