1-Bromo-2-(2-methoxyethoxy)ethane 90% stabilised
:54149-17-6
:
:BrH₂CCH₂OCH₂CH₂OCH₃
:MFCD00000238
:183.05 g/mol
:172 °C (1013 hPa)
:–23 °C
:1,347 g/cm³
:73 °C
:Stabilised with sodium carbonate (Na₂CO₃)
:Refrigerator
:259-000-6
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4540 to 1.4600 (20°C, 589 nm)
- GC:≥88 %
- Stabilizer:0.1 % Sodium carbonate
Frequently Bought Together