Order Entry
Ireland
ContactUsLinkComponent
1-Bromooctane for synthesis, Sigma-Aldrich®
Environment
1-Bromooctane for synthesis, Sigma-Aldrich®
Catalog # 8.01969.0250
Supplier:  Merck
CAS Number:  
Environment
1-Bromooctane for synthesis, Sigma-Aldrich®
Catalog # 8.01969.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Seq:
    2590

Specifications

Specification Test Results

  • Solubility:
    0.002 g/l (20 °C)
  • Melting Point:
    -55 °C
  • Molar Mass:
    193.12 g/mol
  • Boiling Point:
    201 - 202 °C (1013 mbar)
  • Vapor Pressure:
    0.47 hPa (25 °C)
  • Flash Point:
    80 °C
  • Refractive Index:
    1.4503 (25 °C, 589 nm)
  • Density:
    1.11 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :193.13 g/mol
Merck Index:12,06861
Boiling Pt:198…201 °C (1013 hPa)
Melting Pt:–55 °C
Density:1,112 g/cm³ (20 °C)
Storage Temperature:Freezer
EINECS:203-912-9
UN:3082
ADR:9,III
 :M-Clarity™ quality level = MQ200
 :GHS09
CAS Number:111-83-1
Formula:BrH₂C(CH₂)₆CH₃
MDL Number:MFCD00000276