1,4-Dibromobutane 99%
: 1,4-Dibromo butane
:110-52-1
:
:BrH₂C(CH₂)₂CH₂Br
:MFCD00000261
:215.92 g/mol
:197 °C (760 mmHg)
:–20 °C
:1,808 g/cm³
:>112 °C
:Ambient
:203-775-5
:2810
:6.1,III
:GHS05
:GHS06
- Environmentally Preferable:
- Appearance:Clear colorless to yellow-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5176 to 1.5196 (20°C, 589 nm)
- GC:≥98.5 %
- Specific optical rotation:+18° to +23° (20°C, 589 nm) (c=0.5, acetone)
Frequently Bought Together