Order Entry
Ireland
ContactUsLinkComponent
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Toxic
Health
Environment
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Health
Environment
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    0.045 g/l (20 °C)
  • Melting Point:
    -30 °C
  • Molar Mass:
    132.21 g/mol
  • Boiling Point:
    207 °C (1013 hPa)
  • Vapor Pressure:
    0.24 hPa (20 °C)
  • Flash Point:
    71 - 77 °C
  • Refractive Index:
    1.5414 (20 °C, 589 nm)
  • Explosion Limit:
    0.8 - 5 %(V)
  • Density:
    0.97 g/cm3 (20 °C)
  • Ignition Point:
    425 °C DIN 51794
  • Saturation concentration:
    1.3 g/m3 (20 °C) Air
  • Evaporation number:
    190
  • Hazard Class:
    9
  • Hazard Class Description:
    Miscellaneous Hazardous Material

Specification Test Results

Related Information

Frequently Bought Together