Calcium chloride dihydrate 99.0-103.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Calcium Chloride Dihydrate, Calcium dichloride
CAS Number:10035-04-8
:
Formula:CaCl₂·2H₂O
MDL Number:MFCD00149613
Boiling Pt:1600
Melting Pt:176
Density:1.85 g/cm³ (20 °C)
Molecular Weight:147.01 g/mol
Merck Index:13,01660
EINECS:233-140-8
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.0 - 103.0 %
- Appearance of solution:Passes test Ph.Eur.
- Identification A:Passes test Ph.Eur.
- Identification B:Passes test Ph.Eur.
- Solution in water:Passes test
- Al (Aluminium):Passes test Ph.Eur.
- Acidity or alkalinity:Max. 0.002 meq/g
- pH (25°C; 5 %):4.5 - 8.5
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 100 ppm
- Magnesium and alkali metals:Max. 0.5 %
- Not precipitated by (COONH4)2 (as SO4):Max. 0.2 %
- Oxidising substances:Max. 30 ppm
- Total N (Nitrogen):Max. 20 ppm
- NH4 (Ammonium):Max. 50 ppm
- PO4 (Phosphate):Max. 10 ppm
- SO4 (Sulphate):Max. 50 ppm
- As (Arsenic):Max. 1 ppm
- Ba (Barium):Max. 30 ppm
- Cu (Copper):Max. 5 ppm
- Fe (Iron):Max. 3 ppm
- K (Potassium):Max. 100 ppm
- Mg (Magnesium):Max. 100 ppm
- Na (Sodium):Max. 100 ppm
- Sr (Strontium):Max. 0.05 %
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 30L Plastic bucket UN Z (ZPACK13247037) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC22317.230 spec_VWRC22317.230 spec_VWRC22317.260 spec_VWRC22317.260 spec_VWRC22317.297
Product specification
More
Frequently Bought Together