Sodium hydrogen sulphate monohydrate
1.06350.9025
: Merck
:
- Pack type:Fibre carton
- Pk:25 kg
- Seq:4283
- Environmentally Preferable:
: Sodium bisulphate monohydrate
:
:138.08 g/mol
:183
:231-665-7
:3260
:GHS05
:Ambient
:10034-88-5
:NaHSO₄·H₂O
:MFCD00149210
:2.1 g/cm³ (20 °C)
:13,08658