Sodium dihydrogen phosphate dihydrate ≥98.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:13472-35-0
:
Formula:NaH₂PO₄·2H₂O
MDL Number:MFCD00149209
:156.01 g/mol
Merck Index:12,08806
Boiling Pt:< 650 °C (1013 hPa)
Melting Pt:60 °C
Density:1,91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13
Specification Test Results
- Assay:Min. 98.0 %
- Heavy metals (as Pb):Max. 20 ppm
- Cl (Chloride):Max. 50 ppm
- SO4 (Sulphate):Max. 0.05 %
- Fe (Iron):Max. 50 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 33L Plastic bucket UN Z (ZPACK13247037) 6L Plastic bottle for solids DIN 80 (ZPACK13256000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC28011.260 spec_VWRC28011.260 spec_VWRC28011.260 spec_VWRC28011.291 spec_VWRC28011.291
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together