
Sodium carbonate decahydrate 99.0-102.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 27768.298
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay:99.0 - 102.0 %
- Alkali hydroxides and bicarbonates:Passes test
- Solution in water:Passes test
- Heavy metals (as Pb):Max. 5 ppm
- Total N (Nitrogen):Max. 5 ppm
- Total S (as SO4):Max. 30 ppm
- Cl (Chloride):Max. 10 ppm
- PO4 + SiO4 (as SiO2):Max. 20 ppm
- Al (Aluminium):Max. 5 ppm
- As (Arsenic):Max. 5 ppm
- Ca (Calcium):Max. 20 ppm
- Fe (Iron):Max. 2 ppm
- K (Potassium):Max. 50 ppm
- Mg (Magnesium):Max. 3 ppm
Specification Test Results
About this item
:
:286.14 g/mol
Melting Pt:33 °C
Density:1,46 g/cm³ (20 °C)
EINECS:207-838-8
:GHS07
Storage Temperature:Ambient
CAS Number:6132-02-1
Formula:Na₂CO₃·10H₂O
MDL Number:MFCD00149178