Order Entry
Ireland
ContactUsLinkComponent
di-Sodium tetraborate decahydrate, BAKER, J.T.Baker®
di-Sodium tetraborate decahydrate, BAKER, J.T.Baker®
Catalog # 0270.1000
Supplier:  Avantor
CAS Number:  
di-Sodium tetraborate decahydrate, BAKER, J.T.Baker®
Catalog # 0270.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    99.0 - 103.0 %
  • Ammonium (NH4):
    max. 10 ppm
  • Appearance of solution:
    passes test
  • Arsenic (As):
    max. 5 ppm
  • Calcium (Ca):
    max. 100 ppm
  • Heavy Metals (as Pb):
    max. 25 ppm
  • Identification:
    passes test
  • pH:
    9.0 - 9.6
  • Sulfates (as SO4):
    max. 50 ppm

Specification Test Results

About this item

 :
 :381.37 g/mol
Merck Index:13,08662
Boiling Pt:320 °C (1013 hPa)
Melting Pt:75 °C
Density:1,73 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
 :GHS08
CAS Number:1303-96-4
Formula:Na₂B₄O₇·10H₂O
MDL Number:MFCD00149193