di-Sodium tetraborate decahydrate 99.5-105.0% ACS
Catalog # 40114.36
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
CAS Number:1303-96-4
:
Formula:Na₂B₄O₇·10H₂O
MDL Number:MFCD00149193
:381.37 g/mol
Merck Index:14,8590
Boiling Pt:320 °C (1013 hPa)
Melting Pt:75 °C
Density:1,73
Storage Temperature:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
:GHS08