Hydrochloric acid 1 mol/l (1 N), AVS TITRINORM Reag. Ph. Eur., Reag. USP, NF volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1,02 g/cm³
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8,III
:36.46 g/mol
:GHS05
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Conforms to Reag. Ph.Eur.:Passes test
- Titer (20°C; real value 0.2 % accuracy):0.998 - 1.002 mol/l
- Conforms to Reag. USP-NF:Passes test
- NIST traceable:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Number10L Cubitainer UN (ZPACK13260004) 10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 25L Plastic drum for liquids DIN 61 (ZPACK13952500)
Packaging Information
More
spec_VWRC30024.290 spec_VWRC30024.324 spec_VWRC30024.370 spec_VWRC30024.404 spec_VWRC30024.415
Product specification
More
Frequently Bought Together