(±)-2-Octanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:123-96-6
:
Formula:C₈H₁₈O
MDL Number:MFCD00004591
:130.23 g/mol
Merck Index:13,06785
Boiling Pt:180 °C (1013 hPa)
Melting Pt:–39 °C
Density:0,818 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-667-0
UN:1987
ADR:3,III
:M-Clarity™ quality level = MQ200
Flash Pt:55 °C
:GHS07
Specification Test Results
- Solubility:1.12 g/l (25 °C)
- Melting Point:-38 °C
- Molar Mass:130.23 g/mol
- Boiling Point:177 - 182 °C (1013 hPa)
- Vapor Pressure:0.25 hPa (20 °C)
- Flash Point:71 °C
- Refractive Index:1.4234 (20 °C, 589 nm)
- Explosion Limit:0.80 - 7.40 %(V)
- Density:0.82 g/cm3 (20 °C)
- Ignition Point:255 - 260 °C
Specification Test Results
Frequently Bought Together