Order Entry
Ireland
ContactUsLinkComponent
(±)-2-Butanol ≥99% analytical reagent
Flammable
Irritant
(±)-2-Butanol ≥99% analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Irritant
(±)-2-Butanol ≥99% analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.0 %
  • Acidity:
    Max. 0.0005 meq/g
  • Boiling point:
    98.5 - 100.5 °C
  • Density (20/4):
    0.805 - 0.809
  • Evaporation residue:
    Max. 50 ppm
  • Water:
    Max. 0.2 %

Specification Test Results

Frequently Bought Together