Order Entry
Ireland
ContactUsLinkComponent
 
 
 
(±)-2-Butanol ≥98%, GPR RECTAPUR®
 
undefined
 
 
(±)-2-Butanol ≥98%, GPR RECTAPUR®
 

 

  • Assay:
    Min. 98 %
  • Boiling point:
    98.5 - 100.5 °C
  • Density (20/4):
    0.805 - 0.809
  • Evaporation residue:
    Max. 100 ppm

 

Frequently Bought Together