(±)-2-Bromopentane tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107-81-3
:
Formula:H₃CBrCH(CH₂)₂CH₃
MDL Number:MFCD00000160
:151.05 g/mol
Boiling Pt:117…118 °C
Melting Pt:–96 °C
Density:1,220
Flash Pt:20 °C (68 °F)
Storage Temperature:Ambient
EINECS:203-521-3
UN:2343
ADR:3,II
:GHS02
:GHS07
Related Information
Frequently Bought Together