Order Entry
Ireland
ContactUsLinkComponent
(±)-2-Bromopentane 90% tech.
(±)-2-Bromopentane 90% tech.
Catalog # ACRO107231000
CAS Number:  
undefined
(±)-2-Bromopentane 90% tech.
Catalog # ACRO107231000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4393 to 1.4413 (20°C, 589 nm)
  • GC:
    ≥88 % Typical
  • Color scale:
    ≤100 APHA

Specification Test Results

About this item

 :
 :151.05 g/mol
Boiling Pt:117 °C (1013 hPa)
Melting Pt:–95,5 °C
Density:1,22 g/cm³ (20 °C)
Flash Pt:20 °C
Storage Temperature:Ambient
EINECS:203-521-3
UN:2343
ADR:3,II
 :GHS07
 :GHS02
CAS Number:107-81-3
Formula:H₃CBrCH(CH₂)₂CH₃
MDL Number:MFCD00000160