


Nitric acid 68%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01 g/mol
Merck Index:13,06608
Boiling Pt:120,5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8 (5.1),II
:GHS03
:GHS05
:GHS06
Specification Test Results
- Assay:68 - 70 %
- Density (20/4):1.400 - 1.420
- Heavy metals (as Pb):Max. 20 ppm
- Ignition residue (SO4):Max. 50 ppm
- Cl (Chloride):Max. 5 ppm
- SO4 (Sulphate):Max. 5 ppm
- Fe (Iron):Max. 2 ppm
- Pb (Lead):Max. 1 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 25L Plastic drum for liquids DIN 61 (ZPACE8000477) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
spec_VWRC20413.241 spec_VWRC20413.296 spec_VWRC20413.365 spec_VWRC20413.420 spec_VWRC20413.420
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together