Order Entry
Ireland
ContactUsLinkComponent
Hydrofluoric acid 47 - 51%, Environmental Grade Plus
Toxic
Corrosive
Hydrofluoric acid 47 - 51%, Environmental Grade Plus
Catalog # 38746.AP
CAS Number:  
Toxic
Corrosive
Hydrofluoric acid 47 - 51%, Environmental Grade Plus
Catalog # 38746.AP
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

CAS Number:7664-39-3
 :
Formula:HF
MDL Number:MFCD00011346
 :20.01 g/mol
Merck Index:14,4790
Boiling Pt:112 °C
Density:1,16
Storage Temperature:Ambient
EINECS:231-634-8
UN:1790
ADR:8,II
 :GHS06
 :GHS05