Order Entry
Ireland
ContactUsLinkComponent
 
 
 
Hydrofluoric acid 38%
  WAKO639-03295
 :  Wako
 :  
 
 
Hydrofluoric acid 38%
  WAKO639-03295
 :  Wako
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

 

 

 :
 :20.01 g/mol
 :Ambient
 :231-634-8
 :1790
 :8,II
 :GHS06
 :GHS05
 :7664-39-3
 :HF
 :MFCD00011346