Order Entry
Ireland
ContactUsLinkComponent
Hydrofluoric acid 38%
Toxic
Corrosive
Hydrofluoric acid 38%
Catalog # WAKO632-03285
Supplier:  Wako
CAS Number:  
Toxic
Corrosive
Hydrofluoric acid 38%
Catalog # WAKO632-03285
Supplier:  Wako
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :20.01 g/mol
Storage Temperature:Ambient
EINECS:231-634-8
UN:1790
ADR:8,II
 :GHS06
 :GHS05
CAS Number:7664-39-3
Formula:HF
MDL Number:MFCD00011346