Order Entry
Ireland
ContactUsLinkComponent
Potassium chromate 5% in aqueous solution
Health
Environment
Potassium chromate 5% in aqueous solution
Catalog # 160254K
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Environment
Potassium chromate 5% in aqueous solution
Catalog # 160254K
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Assay (W/V):
    4.975 - 5.025 %
  • Cl (Chloride):
    Max. 2 ppm

Specification Test Results

About this item

 :
 :194.19 g/mol
EINECS:232-140-5
UN:3082
ADR:9,III
 :GHS08
 :GHS09
CAS Number:7789-00-6
Formula:K₂CrO₄
MDL Number:MFCD00011368