

Hydrochloric acid fuming 37%, tested according to Ph. Eur., Fluka™
Catalog # 84436-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Vapor Pressure:3.23 psi ( 21.1 °C)
- Identity A:complying
- Identity B:complying
- Assay:35.0 - 39.0 %
- Appearance of the solution:complying
- Non-volatile matter:Max. 0.01 %
- Sulfate (SO₄):Max. 20 ppm
- Free chlorine (Cl):Max. 4 ppm
- Residual solvents:complying
Specification Test Results
About this item
:
Boiling Pt:42 °C (1,013 hPa)
Melting Pt:–35 °C
Density:1,190 g/cm³ (20 °C)
ADR:8,II
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS07
:GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324