Hydrochloric acid ≥32% (by titrimetric analysis), Puriss. p.a., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:42 °C (1,013 hPa)
Melting Pt:–35 °C
Density:1,190 g/cm³ (20 °C)
EINECS:231-595-7
UN:1789
ADR:8,II
:36.46 g/mol
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Assay:32.0 - 35.0 %
- Density (D 20/4):1.150 - 1.170
- Residue on ignition (SO₄):Max. 0.0005 %
- Ammonium (NH₄):Max. 0.0001 %
- Aluminium (Al):Max. 0.05 ppm
- Arsenic (As):Max. 0.01 ppm
- Barium (Ba):Max. 0.02 ppm
- Beryllium (Be):Max. 0.02 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.01 ppm
- Cobalt (Co):Max. 0.01 ppm
- Chromium (Cr):Max. 0.02 ppm
- Copper (Cu):Max. 0.02 ppm
- Iron (Fe):Max. 0.2 ppm
- Germanium (Ge):Max. 0.05 ppm
- Potassium (K):Max. 0.1 ppm
- Lithium (Li):Max. 0.01 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.01 ppm
- Molybdenum (Mo):Max. 0.02 ppm
- Sodium (Na):Max. 0.5 ppm
- Nickel (Ni):Max. 0.02 ppm
- Lead (Pb):Max. 0.02 ppm
- Strontium (Sr):Max. 0.01 ppm
- Titanium (Ti):Max. 0.1 ppm
- Thallium (Tl):Max. 0.05 ppm
- Vanadium (V):Max. 0.01 ppm
- Zinc (Zn):Max. 0.05 ppm
- Zirconium (Zr):Max. 0.1 ppm
- Bromide (Br):Max. 50 ppm
- Free chlorine (Cl):Max. 0.00005 %
- Phosphate (PO₄):Max. 0.5 ppm
- Sulfate (SO₄):Max. 1 ppm
- Sulfite (SO₃):Max. 1 ppm
- Vapor Pressure:3.23 psi ( 21.1 °C)
Specification Test Results
Frequently Bought Together