Hydrochloric acid 25% in water for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:107 °C (760 mmHg)
Melting Pt:–70 °C
Density:1,12 g/cm³
EINECS:231-595-7
UN:1789
ADR:8,II
:36.46 g/mol
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Solution
- Residue after ignition:≤0.0005 %
- Titration with NaOH:≥25.0 %
- Phosphate (PO4):≤0.00005 %
- Sulfate (SO4):≤0.0001 %
- Aluminium (Al):≤0.000005 %
- Barium (Ba):≤0.000002 %
- Bismuth (Bi):≤0.00001 %
- Cadmium (Cd):≤0.000001 %
- Calcium (Ca):≤0.00005 %
- Chromium (Cr):≤0.000002 %
- Cobalt (Co):≤0.000001 %
- Copper (Cu):≤0.000002 %
- Iron (Fe):≤0.00002 %
- Lead (Pb):≤0.000002 %
- Lithium (Li):≤0.000001 %
- Magnesium (Mg):≤0.00001 %
- Manganese (Mn):≤0.000001 %
- Molybdenum (Mo):≤0.000002 %
- Nickel (Ni):≤0.00002 %
- Potassium (K):≤0.00001 %
- Sodium (Na):≤0.00005 %
- Titanium (Ti):≤0.00001 %
- Vanadium (V):≤0.000001 %
- Zirconium (Zr):≤0.00001 %
Specification Test Results
Frequently Bought Together