

Formic acid ≥90.00-90.15%, Supelco®
Catalog # 1.10854.2500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay:90.00-90.15%
- Acetic acid (CH3COOH):0.05% max
Specification Test Results
About this item
:
:46.03 g/mol
ADR:8 (3),
:M-Clarity™ quality level = MQ300
EINECS:200-579-1
UN:1779
:GHS06
:GHS05
Storage Temperature:Ambient
CAS Number:64-18-6
Formula:HCO₂H
MDL Number:MFCD00003297