Formalin 10% (v/v), Histology grade, neutralised (pH 7.0 ± 0.2), J.T.Baker®
: Avantor
:50-00-0
:
:CH₂O
:MFCD00003274
:30.03 g/mol
:200-001-8
:GHS08
:GHS07
:Ambient
:1
- Sodium (Na):61 - 71 mmol/l
- Formaldehyde (HCHO):38 - 42 g/l
- pH at 25°C:6.8 - 7.2
Frequently Bought Together