Order Entry
Ireland
ContactUsLinkComponent
 
Pyridine-[D₅] 99.8 atom % D for NMR spectroscopy
Catalog # ACRO351480075
CAS Number:  
undefined
Pyridine-[D₅] 99.8 atom % D for NMR spectroscopy
Catalog # ACRO351480075
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    7,5 mL
  • Seq:
    2065
  • Environmentally Preferable:
  • Pk Info:
    10 x 0.75 ml

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.03 % (Coulometric)
  • D-Enrichment:
    ≥99.75 %

Specification Test Results

About this item

Synonyms: Pyridine-[²H₅]
 :
 :84.06 g/mol
Boiling Pt:114,4 °C (1013 hPa)
Melting Pt:–41 °C
Density:1,05 g/cm³ (20 °C)
Flash Pt:20 °C
Storage Temperature:Ambient
EINECS:230-720-2
UN:1282
ADR:3,II
 :GHS07
 :GHS02
CAS Number:7291-22-7
Formula:C₅D₅N
MDL Number:MFCD00044639