Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.02 % (Coulometric)
- D-Enrichment:≥99.95 %
- GC:≥98.5 %
- Stabilizer:Copper
Specification Test Results
Frequently Bought Together