0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Acetone-[D6]
Acetone-[D₆]
Acetone-[D₆]
:
Roth Carl
Acetone-[D₆]
:
Roth Carl
:
666-52-4
:
:
D₃CC(O)CD₃
:
MFCD00044635
:
64.03 g/mol
:
55,5 °C (1013 hPa)
:
–95,4 °C
:
0,79 g/cm³ (20 °C)
:
–19 °C (closed cup)
:
Ambient
:
211-563-9
:
1090
:
3,II
:
GHS02
:
GHS07
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
: Merck
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals