Order Entry
Ireland
ContactUsLinkComponent
Acetone-[D₆] 99.9 atom % D for NMR spectroscopy
Acetone-[D₆] 99.9 atom % D for NMR spectroscopy
Catalog # ACRO320620075
CAS Number:  
undefined
Acetone-[D₆] 99.9 atom % D for NMR spectroscopy
Catalog # ACRO320620075
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    7,5 mL
  • Seq:
    2065
  • Pk Info:
    10 x 0.75 ml

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.03 % (Coulometric)
  • D-Enrichment:
    ≥99.85 %

Specification Test Results

About this item

 :
 :64.03 g/mol
Boiling Pt:55,5 °C (1013 hPa)
Melting Pt:–93 °C
Density:0,87 g/cm³
Flash Pt:–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:211-563-9
UN:1090
ADR:3,II
 :GHS07
 :GHS02
CAS Number:666-52-4
Formula:D₃CC(O)CD₃
MDL Number:MFCD00044635