Acetonitrile-[D₃] 99.9 atom % D for NMR spectroscopy
Catalog # ACRO351390100
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.03 % (Coulometric)
- D-Enrichment:≥99.85 %
Specification Test Results
About this item
:
:44.03 g/mol
Boiling Pt:80,7 °C (1013 hPa)
Melting Pt:–46 °C
Density:0,844 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:218-616-5
UN:1648
ADR:3,II
:GHS02
:GHS07
CAS Number:2206-26-0
Formula:D₃CCN
MDL Number:MFCD00001881