Order Entry
Ireland
ContactUsLinkComponent
Diisobutylene, Sigma-Aldrich®
Diisobutylene, Sigma-Aldrich®
  SIAL38180-1L-F
 :  Merck
 :  
undefined
Diisobutylene, Sigma-Aldrich®
  SIAL38180-1L-F
 :  Merck
 :  
  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
 :
 :112.22 g/mol
 :101…102 °C (1013 hPa)
 :–106 °C
 :0,71 g/cm³ (20 °C)
 :–6 °C
 :Ambient
 :246-690-9
 :2050
 :3,II
 :GHS02
 :GHS08
 :GHS09
 :25167-70-8
 :C₈H₁₆
 :MFCD00008855