2-Pentene cis- and trans-mixture ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:109-68-2
:
Formula:C₅H₁₀
MDL Number:MFCD00009384
:70.13 g/mol
Merck Index:14,7123
Boiling Pt:36…38 °C
Melting Pt:–140 °C
Density:0,650
Flash Pt:–45 °C (–49 °F)
Storage Temperature:Refrigerator
EINECS:203-695-0
UN:3295
ADR:3,II
:GHS02
:GHS08
Related Information
Frequently Bought Together