Bottles, narrow neck, Boston type, HDPE, with screw caps, Nalgene®
HDPE, amber with amber PP screw cap.
- Good chemical resistance
- Heat resistant up to +120 °C
- Leakproof
Amber to protect light-sensitive materials. Ideal for storing and shipping liquids.
- Catalog No:
- 215-2559
- 215-2560
- 215-2555
- 215-2556
- 215-2558
- 215-2557
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Frequently Bought Together