
Copper, powder for analysis, Supelco®
: Merck
:7440-50-8
:
:Cu
:MFCD00010965
:63.55 g/mol
:2567 °C (1013 hPa)
:1083,4 °C
:8,94 g/cm³ (20 °C)
:231-159-6
:4.1,III
:M-Clarity™ quality level = MQ300
:3089
:GHS02
:Ambient
- Melting Point:1083 °C
- Molar Mass:63.55 g/mol
- Bulk Density:1290 kg/m3
- Density:8.96 g/cm3 (20 °C)
Frequently Bought Together