VWR® Maximum SF Shoe Covers, Non Skid
VWR® Maximum SF shoe covers are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
- Non skid and anti-static
- Impervious to fluids
- Superior friction coefficient to ensure maximum traction
- Maximum skid resistance on wet and smooth surfaces
- Clean, low linting fabric
- 60 g laminated fabric with PVC sole
- Particle shedding (IEST-RP-CC003.3) level 1 E
- Sterile version (article; 114-9140)
- Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
Tested and manufactured in an ISO certified facility under stringent process controls. Lot controlled and validated through independent laboratory testing.
Length: 359 mm (U*), 292 mm (M), 324 mm (L), 381 mm (XL)
: *U = One size fits all
- Catalog No:
- 113-1369
- 113-1370
- 113-1371
- 113-1372
- 113-1373
- 114-9140
- 113-1374
- 113-1375
- 113-1376
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
Frequently Bought Together