VWR® Advanced SF Cleanroom Overalls
VWR® Advanced SF overalls are manufactured from spunbonded polypropylene fabric, coated with breathable polyethylene film.
- Available with tapered collar or with hood and attached boot covers
- Inset sleeves, zip closure, elasticated cuffs
- Durable fabric, with good breathability and water transmission rate
- Significant fluid and particulate barrier
- Particle shedding: Level II Helmke drum classification
- Soft, cloth-like fabric
Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
- Catalog No:
- 113-1071
- 113-1070
- 113-1061
- 113-1062
- 113-1073
- 113-1063
- 113-1064
- 113-1065
- 113-1066
- 113-1068
- 113-1067
- 113-1069
- 113-1072
- Model:
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
Frequently Bought Together