Order Entry
Ireland
ContactUsLinkComponent
Benedict's reagent for qualitative detection of sugar in urine, Supelco®
Benedict's reagent for qualitative detection of sugar in urine, Supelco®
Catalog # 11945-250ML
Supplier:  Merck
CAS Number:  
Benedict's reagent for qualitative detection of sugar in urine, Supelco®
Catalog # 11945-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
ADR:9,III
 :M-Clarity™ quality level = MQ100
UN:3082
 :GHS09
 :GHS07
Storage Temperature:Ambient
CAS Number:63126-89-6
MDL Number:MFCD00146068