Order Entry
Ireland
ContactUsLinkComponent
 
Patent Blue VF (Disulphine blue), Sigma-Aldrich®
  198218-25G
 :  
undefined
Patent Blue VF (Disulphine blue), Sigma-Aldrich®
  198218-25G
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Wavelength:
    635 - 641 nm
  • Extinction coefficient:
    ≥50000
  • Carbon:
    28.60 - 75.00 %
  • Nitrogen:
    2.40 - 7.40 %
  • Solubility (Colour):
    Dark blue to very dark blue and dark blue-green to very dark blue-green

 

 

 :
 :566.67 g/mol
 :12,09161
 :290 °C
 :M-Clarity™ quality level = MQ200
 :Room Temperature
 :204-934-1
 :129-17-9
 :C₂₇H₃₁N₂NaO₆S₂
 :MFCD00012119